Diamond Thermo RSH 10 to 50μL GC Autosampler Syringes
Actual product may vary
Thermo Scientific
Diamond Thermo RSH 10 to 50μL GC Autosampler Syringes
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
Have Questions?
Catalog NumberDiameter (Metric) Inner NeedleIncludes
SGE03556300.24 mmUltra Smooth Coating
Catalog number SGE0355630
Price (MYR)
819.00
Each
Diameter (Metric) Inner Needle:
0.24 mm
Includes:
Ultra Smooth Coating
Price (MYR)
819.00
Each
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
DescriptionNeedle coating: Ultra Smooth
Needle Gauge23 G
Diameter (English) Inner Needle0.009 in.
Diameter (Metric) Inner Needle0.24 mm
Diameter (English) Outer Needle0.024 in.
Diameter (Metric) Outer Needle0.63 mm
For Use With (Application)Gas chromatography
IncludesUltra Smooth Coating
Length (English) Needle2.24 in.
Product TypeGC Autosampler Syringe
QuantityEa.
Volume (Metric)10 to 50 μL
Needle Point StyleCone
For Use With (Equipment)Diamond Thermo RSH
Unit SizeEach