Cleanroom Pens
Cleanroom Pens
Actual product may vary
Cleanroom Pens
Cleanroom Pens
Micronova
Cleanroom Pens
These low-sodium, indelible ink ballpoint pens feature a unique polypropylene barrel that limits particle generation and holds up to IPA, bleach and phenol wipe downs. Available irradiated, capped or click versions, and with black, blue or red ink.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
MICPEN-02Blue
MICPEN-01Black
Catalog number MICPEN-02
Price (MYR)
218.00
Pack
Add to cart
Color:
Blue
Price (MYR)
218.00
Pack
Add to cart
Low-sodium ink
The Micronova Cleanroom Ballpoint Pens, with low sodium ink and low particulate barrels, are designed for documentation and labeling in cleanrooms and aseptic environments. Gamma irradiated pens are suitable for Aseptic ISO 5 and other clean production areas.
  • Ball point cleanroom pens with indelible, low sodium ink
  • Designed for documentation and labeling in cleanrooms
  • White, high-density polypropylene barrel
  • Holds up to IPA, bleach and phenol wipe downs
  • Capped or Retractable options available
  • Black, blue or red ink
  • Available gamma irradiated for aseptic environments
Specifications
MaterialHDPE
ColorBlue
DescriptionCapped
Length (English)5.75 in.
AutoclavableNot Autoclavable
DisposableReusable
For Use With (Application)Cleanroom and Aseptic Environment Documentation and Labeling
Product TypeCleanroom Pen
SterilityNon-sterile
Tip Style0.7 mm Point Tip
Unit SizePack