FreeZone Benchtop Shell Freezer, 230 V, Schuko Plug and Receptacle
Actual product may vary
Labconco
FreeZone Benchtop Shell Freezer, 230 V, Schuko Plug and Receptacle
Provides a means to freeze a thin coating of sample on inside walls of flasks, shell frozen samples have greater surface area resulting in enhanced freeze drying
Have Questions?
Catalog NumberCapacity (Metric)
LBC794903012 L
Catalog number LBC7949030
Price (MYR)
42,787.00
Each
Capacity (Metric):
12 L
Price (MYR)
42,787.00
Each
  • 1/2 hp refrigeration system prefreezes samples to -40 °C
  • Stainless-steel bath with two rollers to rotate flasks in low-temperature heat transfer solution
    • Produces uniform thin coating of sample on container interior wall
      • Ensures efficient vapor flow from sample to collector during lyophilization
  • Accepts Fast-Freeze flasks up to 1200 mL in size
  • Does not contain ozone-depleting chlorofluorocarbons (CFCs)
  • LED display of bath temperature from -20 to -42 °C in predetermined increments
  • Left-mounted retractable bath hose facilitates cleaning of bath
  • Fits on countertop or lab cart
  • CE conformity (230 V, 50 Hz model)
  • White powder-coated steel cabinet with brushed stainless-steel front and blue accents
  • Stainless-steel bath with urethane foam insulation and white HDPE cover
Specifications:
  • Shipping weight: 44 kg (96 lb.)
  • Cabinet dimensions (L x W x H): 56.1 x 38.4 x 50 cm (22.1 x 15.1 x 19.7 in.)
  • Bath dimensions (D x W x H): 30.5 x 14 x 16.5 cm (12 x 5.5 x 6.5 in.)
Specifications
TypeFast-Freeze Flask Shell Freezer
Capacity (Metric)12 L
Capacity (English)0.42 feet3
Temperature Range (Metric)-40°C
Temperature Range (English)-40°F
MaterialStainless Steel
Certifications/ComplianceCE
For Use With (Equipment)Freeze dryers
Dimensions (L x W x H) Exterior56.1 x 36.8 x 50 cm
Depth (English)22.1 in.
Depth (Metric)56.1 cm
Width (English)14.5 in.
Width (Metric)36.8 cm
Height (Metric)50 cm
Length (Metric)56.1 cm
Amperage3 A
Plug TypeSchuko
Voltage230 V
Frequency50 Hz
Horsepower1/2 HP
Shipping Weight (English)96 lb.
Shipping Weight (Metric)44 kg
Weight (English)95 lb.
Weight (Metric)43 kg
Unit SizeEach