FlaskScrubber Starter Kit
Actual product may vary
Labconco
FlaskScrubber Starter Kit
Outfit a FlaskScrubber to wash a variety of narrow-mouth labware.
Have Questions?
Catalog NumberNo. of Pins
LBC457720048
Catalog number LBC4577200
Price (MYR)
21,257.00
Each
No. of Pins:
48
Price (MYR)
21,257.00
Each
  • Use to wash a variety of narrow-mouth labware
  • Includes 24-Place Pipette Insert (4575400), Graduated Cylinder Insert (4574600), Utensil Basket with Cover (4573100), 48-Pin Insert (4574200), and Standard Upper Rack (4668700)
Specifications
MaterialStainless Steel
Product TypeStarter Kit
ColorSilver
No. of Pins48
Rack TypeSpindle
For Use With (Application)Washing Glassware
For Use With (Equipment)FlaskScrubber Glassware Washer
Includes24-Place Pipette Insert (4575400), Graduated Cylinder Insert (4574600), Utensil Basket with Cover (4573100), 48-Pin Insert (4574200), and Standard Upper Rack (4668700)
Dimensions (L x W x H)17.7 x 8.9 x 5.1 in.
Weight (English) Shipping20 lb.
Weight (Metric) Shipping9.1 kg
Unit SizeEach