Kimtech™ Pure G5 Latex 12 in. Cleanroom Gloves
Kimtech™ Pure G5 Latex 12 in. Cleanroom Gloves
Actual product may vary
Kimberly-Clark
Kimtech™ Pure G5 Latex 12 in. Cleanroom Gloves
G5 non-sterile 12 in. latex gloves provide enhanced tactile sensitivity and performance. Recommended for Class 100/ISO 5 or higher critical environments.
Have Questions?
Catalog NumberSize
KLCHC2211Small
Catalog number KLCHC2211
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
Small
  • Low in natural rubber proteins
  • Textured palm and fingers
  • Beaded cuff adds strength and makes donning easier
  • Ambidextrous
  • Thickness: 8.3 mil
  • Length: 12 in. (30 cm)
Specifications
Certifications/ComplianceASTM D3578
Certifications/Compliance (Europe)Cat III according to (EU) 2016/425, EN ISO 374-1:2016, EN 374-4:2014, EN ISO 374-5:2016
Cleanroom ClassClass 100 (ISO 5)
AQL1.5
ColorNatural
Exterior FinishNo Texture
HandAmbidextrous
Latex FreeNo
MaterialLatex
PowderedPowder Free
SizeSmall
SterilityNon-sterile
Thickness (English)8 mil
Thickness (Metric)0.20 mm
Model56809
SeriesKimtech™ Pure G5
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCS