ColorClear
For Use With (Equipment)4 Block Systems
MaterialAcrylic
TypeSafety Cover
For Use With (Application)For maximum visibility whilst preventing accidental touching of a hot block or contamination of samples from splashes
Height (Metric)85 mm
Unit SizeEach