Clobetasol propionate
Thermo Scientific Chemicals
Clobetasol propionate
Have Questions?
Catalog NumberQuantity
FSA4668200505 g
Catalog number FSA466820050
Price (MYR)
1,367.00
Each
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,367.00
Each
Specifications
Chemical Name or MaterialCloβsol propionate
CAS25122-46-7
Quantity5 g
SolubilitySolubility aq. soln.: insoluble.
EINECS Number246-634-3