Poly(methylhydrosiloxane), trimethylsilyl terminated
Poly(methylhydrosiloxane), trimethylsilyl terminated
Poly(methylhydrosiloxane), trimethylsilyl terminated
Thermo Scientific Chemicals
Poly(methylhydrosiloxane), trimethylsilyl terminated
CAS: 63148-57-2 | (CH4OSi)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number FSA440100250
Price (MYR)
481.00
Each
Quantity:
25 mL
Request bulk or custom format
Price (MYR)
481.00
Each
Specifications
Chemical Name or MaterialPoly(methylhydrosiloxane)
CAS63148-57-2
Density1.0060g/mL
Flash Point204°C
Infrared SpectrumAuthentic