Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
FSA424005000500 mLGlass Bottle
Catalog number FSA424005000
Price (MYR)
2,437.00
Bottle
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,437.00
Bottle
Specifications
Chemical Name or MaterialNitric acid
ColorColorless
CAS7697-37-2
Melting Point-42.0°C
Boiling Point122.0°C