Soda lime, pure, granular, with indicator white to violet
Soda lime, pure, granular, with indicator white to violet
Soda lime, pure, granular, with indicator white to violet
Thermo Scientific Chemicals
Soda lime, pure, granular, with indicator white to violet
CAS: 8006-28-8 | CaHNaO2 | 96.074 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass Bottle
Catalog number FSA388352500
Price (MYR)
341.00
Bottle
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
341.00
Bottle
Specifications
Chemical Name or MaterialSoda lime
ColorBeige-Gray
Physical FormGranular
CAS8006-28-8
Health Hazard 1GHS Signal Word: Danger
View more
Gas Adsorbent, Indicator
RUO – Research Use Only