N,N-Diisopropylethylamine, 99.5+%
N,N-Diisopropylethylamine, 99.5+%
N,N-Diisopropylethylamine, 99.5+%
Thermo Scientific Chemicals
N,N-Diisopropylethylamine, 99.5+%
CAS: 7087-68-5 | C8H19N | 129.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
50 mL
100 mL
1 L
2.5 L
Packaging:
View all
Glass bottle
Catalog number FSA367840250
Price (MYR)
247.00
Bottle
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
247.00
Bottle
Specifications
Chemical Name or MaterialN, N-Diisopropylethylamine
Name Note99.5+%
CAS7087-68-5
CAS Min %99.5
Health Hazard 1GHS Signal Word: Danger