Silica gel, for drying purposes, non-toxic grade, 3-6 mm
Silica gel, for drying purposes, non-toxic grade, 3-6 mm
Silica gel, for drying purposes, non-toxic grade, 3-6 mm
Thermo Scientific Chemicals
Silica gel, for drying purposes, non-toxic grade, 3-6 mm
Silica gel, O2Si, CAS Number-7631-86-9, Silicon dioxide, 25kg, 2.0% max. (140 deg.C, 4 hrs), Adsorption capacity (relative humidity 40%): ca 20%, 620 to 800g/L, CHEBI:30563, White to Yellow, 231-545-4, 60.08, Non-toxic, VYPSYNLAJGMNEJ-UHFFFAOYSA-N, dioxosilane, 15, 8632, MFCD00011232 | CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
1 kg
Packaging:
View all
Fiber Drum
Catalog number FSA357400250
Price (MYR)
4,940.00
Each
Quantity:
25 kg
Packaging:
Fiber Drum
Request bulk or custom format
Price (MYR)
4,940.00
Each
Specifications
Chemical Name or MaterialSilica gel
ColorWhite to Yellow
Physical FormSolid
pH5.5 to 8
Name Note3-6mm
View more
Drying agent, For column chromatography
RUO – Research Use Only