Tetramethylammonium chloride, 98+%
Tetramethylammonium chloride, 98+%
Tetramethylammonium chloride, 98+%
Thermo Scientific Chemicals
Tetramethylammonium chloride, 98+%
CAS: 75-57-0 | C4H12ClN | 109.6 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA138332500250 gPlastic bottle
Catalog number FSA138332500
Price (MYR)
474.00
Bottle
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
474.00
Bottle
Specifications
Chemical Name or MaterialTetramethylammonium chloride
Name Note98+%
CAS75-57-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes damage to organs if swallowed.
Toxic to aquatic life with long lasting effects.
View more
Phase transfer catalyst
RUO – Research Use Only