n-Octane, 99+%, extra pure
n-Octane, 99+%, extra pure
n-Octane, 99+%, extra pure
Thermo Scientific Chemicals
n-Octane, 99+%, extra pure
CAS: 111-65-9 | C8H18 | 114.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
2 mL
500 mL
Packaging:
Glass Bottle
Catalog number FSA129370025
Price (MYR)
2,597.00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,597.00
Each
Specifications
Chemical Name or Materialn-Octane
CAS111-65-9
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0