Methanesulfonyl chloride, 99.5%
Methanesulfonyl chloride, 99.5%
Methanesulfonyl chloride, 99.5%
Thermo Scientific Chemicals
Methanesulfonyl chloride, 99.5%
CAS: 124-63-0 | CH3ClO2S | 114.54 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA125642500250 mLGlass bottle
Catalog number FSA125642500
Price (MYR)
467.00
Bottle
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
467.00
Bottle
Specifications
Chemical Name or MaterialMethanesulfonyl chloride
CAS124-63-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic in contact with skin.
May cause an allergic skin reaction.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
May cause respiratory irritation.
May be corrosive
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at