Dioctyl sulfosuccinate, sodium salt, 96%
Dioctyl sulfosuccinate, sodium salt, 96%
Dioctyl sulfosuccinate, sodium salt, 96%
Thermo Scientific Chemicals
Dioctyl sulfosuccinate, sodium salt, 96%
CAS: 577-11-7 | C20H37NaO7S | 444.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA117101000
Price (MYR)
378.00
Bottle
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
378.00
Bottle
Specifications
Chemical Name or MaterialDioctyl sulfosuccinate, sodium salt
Acidity2.5 max. (on solids basis)
CAS577-11-7
ColorWhite
Infrared SpectrumAuthentic