Benzyl benzoate, 99+%
Benzyl benzoate, 99+%
Benzyl benzoate, 99+%
Thermo Scientific Chemicals
Benzyl benzoate, 99+%
CAS: 120-51-4 | C14H12O2 | 212.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
5 L
Packaging:
View all
Glass bottle
Catalog number FSA105860010
Price (MYR)
669.00
Bottle
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
669.00
Bottle
Specifications
Chemical Name or MaterialBenzyl benzoate
Acidity0.3mg KOH/g max.
CAS120-51-4
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning