Barnstead™ 1/2 Size B-Pure™ Filters
Barnstead™ 1/2 Size B-Pure™ Filters
Actual product may vary
Barnstead™ 1/2 Size B-Pure™ Filters
Pre-treat your feed water with Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Filters, available in varying pore sizes.
Have Questions?
Catalog NumberDescription
FL583X50.45µm Final Filter for 1/2 Size B-Pure Filter Holder
Catalog number FL583X5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
0.45µm Final Filter for 1/2 Size B-Pure Filter Holder
Pre-treat your feed water with Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Filters. In addition to pre-treating feed water, the filters can act as a final filter to ensure consistently pure water. Compatible with B-Pure 1/2 size filter holders, these filters are available in varying pore sizes to meet all of your pre-filtration needs.
  • Large surface area for long life
  • Flow rates of up to 2L/min.
Specifications
Description0.45µm Final Filter for 1/2 Size B-Pure Filter Holder
Feedwater SourcePretreated
Flow RateUp to 2 L/min.
Pore Size0.45 μm
For Use With (Equipment)1/2 Size B-Pure Filter Holders
TypeFinal Filter
Length (English)10 in.
Length (Metric)25.4 cm
Diameter (English)2.5 in.
Diameter (Metric)6.5 cm
Unit SizeEA