Nunc™ Lab-Tek™ II Chamber Slide™ System
Actual product may vary
Nunc™ Lab-Tek™ II Chamber Slide™ System
Nunc™ Lab-Tek™ II Chamber Slide™ System
Fisherbrand
Nunc™ Lab-Tek™ II Chamber Slide™ System
Grow cells directly on slides with the Nunc Lab-Tek II Chamber Slide System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWells
FKS162606618
FKS162506614
Catalog number FKS16260661
Price (MYR)
2,997.00
Pack
Wells:
8
Price (MYR)
2,997.00
Pack
Grow, stain, and examine cells directly on the Thermo Scientific Nunc Lab-Tek II Chamber Slide, which consists of a removable polystyrene media chamber attached to a standard glass slide. The slide surface is specially treated for adherent cell culture. Available in 1-, 2-, 4-, and 8-well formats.
Features
  • Slide surface that is specially treated with a unique RS wash to ensure consistent cell growth
  • Tabbed polystyrene lid that allows gas exchange with minimal evaporation
  • Removal tool that separates media chamber from glass slide
  • Biocompatible adhesive hydrophobic well border printed on slide
  • White frosted writing area
Includes: chamber removal tool
Additional information
  • Can be purchased in case of 96 or pack of 16
  • Shipped sterile in convenient stackable incubation trays
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Wells8
Culture Area0.7 cm2
Lid MaterialPolystyrene
Description8-well Chamber Slide w/ removable wells
Surface TreatmentCell Culture
MaterialRS Glass
Volume (Metric) Working0.2 to 0.5 mL
IncludesClear polystyrene lid with handles on the top and side for aseptic manipulations
No. per Pack16
Dimensions (LxW)74 x 25 mm
SterilitySterile
Closure ColorClear
Quantity1 Ea.
Unit SizePack