Manufactured from borosilicate glass. Scientific Glass Labs™ Borosilicate Glass Powder Funnel is less likely to clog or cake when funneling powdery substances.
Have Questions?
Catalog Number
Diameter (Metric)Top
Diameter (Metric) Stem
Length (Metric) Stem
FKS12903601
100mm
22mm
25mm
Catalog number FKS12903601
Price (MYR)
102.00
Each
Diameter (Metric)Top:
100mm
Diameter (Metric) Stem:
22mm
Length (Metric) Stem:
25mm
Price (MYR)
102.00
Each
Manufactured from borosilicate glass.
Short stem funnel
Less likely to clog or cake when funneling powdery substances