Round Glass Narrow Mouth Alpha Bottles
Actual product may vary
Fisherbrand
Round Glass Narrow Mouth Alpha Bottles
Store, pour or share your laboratory liquids using Round Glass Narrow Mouth Alpha Bottles. Available in clear or amber glass, these round bottles are an integral tool in the laboratory.
Have Questions?
Catalog NumberCapacity (Metric)Color
FKS12840904250 mLClear
Catalog number FKS12840904
Price (MYR)
353.00
Pack
Capacity (Metric):
250 mL
Color:
Clear
Price (MYR)
353.00
Pack
  • Narrow neck
  • Borosilicate glass, round
  • Amber or clear glass
Specifications
Capacity (Metric)250 mL
ColorClear
MaterialSoda Lime Glass
No. per Pack60Pack
Quantity60Pack
MouthNarrow
ShapeRound
Height (Metric)143 mm
Product TypeAlpha Bottle
Diameter (Metric)62 mm
Unit SizePack