Whatman™ Plastic Zipper Seal Storage Bag
Actual product may vary
Fisherbrand
Whatman™ Plastic Zipper Seal Storage Bag
Zipper storage bag for 903 Protein Saver Cards
Have Questions?
Catalog NumberMaterial
FKS11969134Plastic
Catalog number FKS11969134
Price (MYR)
622.00
Pack
Material:
Plastic
Price (MYR)
622.00
Pack
  • Accommodates cards up to W x H: 95 x 130 mm
  • W x H: 4 x 6 in. (102 x 152 mm)
  • Clear flexible plastic
  • Pack of 100
Specifications
Closure TypeZip Seal
ColorClear
For Use With (Application)Storing and archiving Whatman Protein Saver card samples.
MaterialPlastic
No. per Pack100
Quantity100
Width (English)4 in.
Height (English)6 in.
Height (Metric)152 mm
Product TypeZip-seal Storage Bag
Width (Metric)102 mm
Unit SizePack