Search Thermo Fisher Scientific
| Catalog Number | Height (Metric) | No. of Shelves |
|---|---|---|
| FKS11917874 | 45.7 cm | 4 Removable Adjustable and Vented |
| FIS08-642-23A | 17.8 cm | 2 Removable Adjustable and Vented |
| FIS08-642-23B | 30.5 cm | 3 Removable Adjustable and Vented |
Dust- and moisture-free storage in refrigerators, freezers and on benchtops