Microcap™ Disposable Micropipets
Actual product may vary
Fisherbrand
Microcap™ Disposable Micropipets
Fast, safe, clean, and accurate way to transfer liquids in micro quantities
Have Questions?
Catalog NumberVolume (Metric)Length (Metric)
FKS118886610.5 μL32 mm
Catalog number FKS11888661
Price (MYR)
530.00
Pack
Volume (Metric):
0.5 μL
Length (Metric):
32 mm
Price (MYR)
530.00
Pack
  • Precision-bore glass capillary tubes
  • Disposable microcaps
  • With spotting holder and bulb dispenser
Specifications
Diameter (English) Inner0.0039 in.
Diameter (English) Outer0.021 in.
Diameter (Metric) Inner0.09 mm
Diameter (Metric) Outer0.53 mm
MaterialGlass
For Use With (Application)For handling dangerous materials, fast, safe, clean and accurate way to transfer liquids in micro quantities
IncludesSpotting holder and bulb dispenser
Length (English)1.25 in.
Volume (Metric)0.5 μL
Length (Metric)32 mm
TypeMicropipet