Western Blotting Filter Papers, 0.83 mm thick, 8 x 10.5 cm
Western Blotting Filter Papers, 0.83 mm thick, 8 x 10.5 cm
Actual product may vary
Western Blotting Filter Papers, 0.83 mm thick, 8 x 10.5 cm
Western Blotting Filter Papers, 0.83 mm thick, 8 x 10.5 cm
Western Blotting Filter Papers, 0.83 mm thick, 8 x 10.5 cm
Western Blotting Filter Papers, 0.83 mm thick, 8 x 10.5 cm
Western Blotting Filter Papers, 0.83 mm thick, 8 x 10.5 cm
Fisherbrand
Western Blotting Filter Papers, 0.83 mm thick, 8 x 10.5 cm
Pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins from polyacrylamide gels to PVDF, nitrocellulose, or other membranes.
Have Questions?
Catalog NumberQuantity
FKS11814131100 Sheets
Catalog number FKS11814131
Price (MYR)
1,091.00
PK
Add to cart
Quantity:
100 Sheets
Price (MYR)
1,091.00
PK
Add to cart
Thermo Scientific™ Pierce Western Blotting Filter Papers are pre-cut cotton sheets for wet or semi-dry, passive or electrophoretic transfer of proteins from polyacrylamide gels (SDS-PAGE) to PVDF, nitrocellulose, or other membranes.
Sheets of blotting paper are necessary components of transfer sandwiches and cassettes that typically must be assembled for various kinds of protein or nucleic acid, gel-to-membrane, transfer protocols. High-quality Pierce Western Blotting Filter Papers are available as standard-thickness and extra-thick sheets that are pre-cut for convenience with mini gels.
Highlights:
  • High-quality – clean cotton cellulose fiber paper manufactured with additive-free ultrapure water to eliminate sources of background signal and artifacts
  • Easy to use – pre-cut filter paper sheets in several convenient sizes for use with most mini gel sizes, tank transfer cassettes and semi-dry blotters
  • Two thicknesses – choose standard-thickness paper for traditional procedures or extra-thick filter paper for high capacity blotting or to eliminate using multiple sheets
  • Validated – tested and recommmend for use with protein methods, including the Thermo Scientific Pierce Fast Semi-Dry Blotter (Part No. 88217)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce Western Blotting Filter Paper, 8cm x 10.5cm
FormatSheet
MaterialCotton
Filtration Area8 x 10.5 cm
Quantity100 Sheets
Thickness (Metric)0.83 mm
TypeWestern Blotting Filter Paper
Dimensions (L x W)8 x 10.5 cm
Unit SizePK
Contents & Storage
Store at room temperature.