Retort Stand Clamps
Actual product may vary
Fisherbrand
Retort Stand Clamps
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
Have Questions?
Catalog NumberSize (Metric) Grip
FKS1178537685 mm
Catalog number FKS11785376
Price (MYR)
192.00
Each
Size (Metric) Grip:
85 mm
Price (MYR)
192.00
Each
  • Design ensures a firm grip at all angles of opening
  • Cork liners for the jaws are well secured
Specifications
TypeRetort Clamp
MaterialAluminum
IncludesCork-lined Jaws
For Use With (Equipment)For use with 85mm diameter
Size (Metric) Grip85 mm
Jaw TypeFour Interlocking
Unit SizeEach