Mini Plate Spinner Centrifuge
Actual product may vary
Fisherbrand
Mini Plate Spinner Centrifuge
Specifically designed for quick and easy centrifugation of samples in PCR plates or microplates
Have Questions?
Catalog NumberElectrical Requirements
FKS11766427230 V
Catalog number FKS11766427
Price (MYR)
-
Electrical Requirements:
230 V
Specifically designed for quick and easy centrifugation of samples in PCR plates or microplates
  • Speed: 2500rpm
  • Capacity: 2 plates
  • Weight: 1.5kg (3.5 lbs)
  • Dimensions: 6 in.×6 in.×7 in.
  • Exceptionally small footprint
  • Rotor designed for rapid start to stop
Two standard microplates are loaded through an opening in the top of the unit. Sealed plates are loaded vertically and surface tension keeps samples in place so there is no danger of spilling samples. Once the lid is closed, the rotor quickly accelerates to 2,500 rpm. Pressing the open button engages the friction brake and brings the rotor to a rapid stop. For temperature sensitive applications, the Plate Spinner may be used in a cold room.
Specifications
Capacity2 Plates
Max. Speed2,500 rpm
Includes2 year warranty
Electrical Requirements230 V
Net Weight3.5 lb. (1.58 kg)
Voltage230 V
Product TypeMini Plate
Unit SizeEach