Viton™ L/S™ 2-Stop Pump Tubing
Viton™ L/S™ 2-Stop Pump Tubing
Actual product may vary
Fisherbrand
Viton™ L/S™ 2-Stop Pump Tubing
Good for food and lab applications
Have Questions?
Catalog NumberTubing Size
FKS1175015916
Catalog number FKS11750159
Price (MYR)
3,998.00
PK
Add to cart
Tubing Size:
16
Price (MYR)
3,998.00
PK
Add to cart
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work
  • Good chemical resistance, opaque black
  • For use with Masterflex™ multichannel pump heads
Specifications
Certifications/ComplianceFDA-Compliant
Diameter (Metric) Inner3.1 mm
MaterialViton
Overall Length (English)1.25 ft.
Overall Length (Metric)0.38 m
Min. Operating Temperature (Metric)0°C
Packaging Quantity1 / Pk.
Tubing Size16
Flow Rate460 mL/min
Hose Barb1/8 in.
Vacuum Level660 mm Hg
Max. Operating Temperature (Metric)150°C
Product TypeStopped Pump Tubing
Unit SizePK