Modular Coliwasa
Actual product may vary
Fisherbrand
Modular Coliwasa
Uniquely designed top spin bar and bottom valve plate mechanism provides a positive seal for sample retention along with greater control for releasing the sample
Have Questions?
Catalog NumberMaterial
FKS11724129PVC
Catalog number FKS11724129
Price (MYR)
7,183.00
Each
Material:
PVC
Price (MYR)
7,183.00
Each
Uniquely designed top spin bar and bottom valve plate mechanism provides a positive seal for sample retention along with greater control for releasing the sample
Specifications
Product TypeModular Coliwasa
MaterialPVC
IncludesWith a cleaning brush.
Diameter (Metric) Inner48 mm
Length (Metric)120 cm
Unit SizeEach