Stainless Steel Micro Spoons
Actual product may vary
Fisherbrand
Stainless Steel Micro Spoons
Designed using high quality components these micro spoons feature a natural quality and bright finish.
Have Questions?
Catalog NumberLength (Metric)
FKS11593482150 mm
Catalog number FKS11593482
Price (MYR)
220.00
Pack
Length (Metric):
150 mm
Price (MYR)
220.00
Pack
  • Designed using high quality components these micro spoons feature a natural quality and bright finish
  • Laboratory Micro Spoon, stainless steel 304 spec 18/8 non-magnetic, bright finish
  • Length = 100mm long
  • Spoon measurement 8mm wide x 6mm long x 1.5mm deep
  • Centre barrel diameter 1.60mm - 'micro'
  • Spoon capacity [liquid] = 0.2ml
  • Supplied in standard packs of 10
Specifications
Quantity5 Pk.
Capacity (Metric)150 mL
Product TypeSpoon
Width (Metric)4 mm
Diameter (Metric) Outer3.6 mm
Dimensions (L x W)150 x 4 mm
MaterialStainless Steel
Length (Metric)150 mm
Unit SizePack