Pyrex™ Glass Graduated Cylinder, Class A, Works Certified
Actual product may vary
Fisherbrand
Pyrex™ Glass Graduated Cylinder, Class A, Works Certified
Manufactured from chemically resistant borosilicate glass. Pyrex™ Glass Graduated Cylinders, Class A, Works Certified are individually tested and supplied with certificate showing exact volume contained at 5 points on scale.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS1139354410 mL0.2 mL
Catalog number FKS11393544
Price (MYR)
1,140.00
Pack
Capacity (Metric):
10 mL
Graduations:
0.2 mL
Price (MYR)
1,140.00
Pack
Manufactured from chemically resistant borosilicate glass. Pyrex™ Glass Graduated Cylinders, Class A, Works Certified are individually tested and supplied with certificate showing exact volume contained at 5 points on scale.
  • Features: sturdy hexagonal base, white ceramic markings, wide range of capacities
  • Class A: conformity certified to German calibration regulations, imprinted with conformity symbol "H" 
  • Batch certification: inscribed with individual serial number for traceability to national standards
  • Complies: ISO 4788
Specifications
MaterialBorosilicate Glass
Capacity (Metric)10 mL
IncludesCertificate Showing Exact Volume Contained at 5 Points on the Scale
ScaleWhite enamel
Graduations0.2 mL
Base TypeHexagonal
Scale ColorWhite
ShapeRound
ClassClass A
Height (English)5.51 in.
Height (Metric)140 mm
Certifications/ComplianceBS 604, DIN 12680, ISO 4788
Tolerance±0.1 mL
Product TypeGraduated Cylinder
Unit SizePack