Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
Actual product may vary
Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
Fisher Catalogue
Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
Conduct high-speed centrifugation of aggressive chemicals with these tubes, suited for chloroform and phenol extractions in refrigerated machines up to 50,000 x g.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric)
FKS1038483150 mL28.8 mm
3114-0050
also known as NLG3114-0050
50 mL28.8 mm
Catalog number FKS10384831
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
50 mL
Diameter (Metric):
28.8 mm
Conduct high-speed centrifugation of aggressive chemicals with Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™, specifically suited for chloroform and phenol extractions.
Features:
  • Withstand temperatures from -100° to +150°C (-148° to +302°F)
  • Leakproof† closures prevent material loss under ordinary use for most spins <10,000 × g
  • When equipped with a Nalgene sealing cap assemblies (sold separately), 30mL and 50mL tubes can be spun up to 50,000 x g
  • Must be filled to at least 100% total capacity for proper performance. 
  • Autoclavable/Biohazard/Leakproof† when used with supplied Nalgene closure /PTFE FEP
Ordering Information:
Optional sealing cap assemblies available — DS3131-0020 (for 30mL tube) and DS3131-0024 (for 50mL tube). Cat. No. 3114-0050 has a brim capacity of only 42mL but is designed to fit standard rotor.
Includes:
Screw caps. Additional sealing cap assembly available separately (DS3131-series).
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Closure MaterialETFE
Capacity (Metric)50 mL
MaterialFEP
No. per Case10
No. per Pack2
SterilityNon-sterile
AutoclavableAutoclavable
ShapeRound
Bottom ShapeRound
Closure TypeScrew Cap
Writing AreaNo
Diameter (Metric)28.8 mm
GraduatedNo
Product TypeCentrifuge Tube
Max. RCF50,000 x g (with sealing cap assembly)
Unit SizePK