Pipet Support Rack
Pipet Support Rack
Actual product may vary
Pipet Support Rack
Pipet Support Rack
Fisherbrand
Pipet Support Rack
Sturdy rack holds 50 pipets in a vertical 5 x 10 arrangement for safe, convenient storage
Have Questions?
Catalog NumberPositions
FKS1022314350
Catalog number FKS10223143
Price (MYR)
610.00
EA
Add to cart
Positions:
50
Price (MYR)
610.00
EA
Add to cart
Sturdy rack holds 50 pipets in a vertical 5 x 10 arrangement for safe, convenient storage
  • Top and middle decks have 16mm diameter holes; Bottom deck holes are tapered from 16 mm to 6 mm diameter to prevent tips from touching benchtop
  • End plates have handles for easy carrying
  • Rugged polypropylene
  • Steam autoclavable at 121°C (250°F)
Specifications
Product TypePipet Rack
Array10 x 5
AutoclavableAutoclavable
ColorTranslucent
HoldsPipets
MaterialPolypropylene
Positions50
ShapeRectangular
Shelves3 Tiers
StackableNo
SterilityNon-sterile
Dimensions (L x W x H)8.37 x 4.5 x 8.87 in. (21.25 x 11.43 x 22.52cm)
Length (Metric)21.25 cm
Length (English)8.37 in.
Width (Metric)11.43 cm
Width (English)4.5 in.
Height (Metric)22.52 cm
Height (English)8.87 in.
Unit SizeEA