Rack Packed Tubes
Actual product may vary
Fisherbrand
Rack Packed Tubes
Compatible with standard microplates and suitable for working with multi-channel pipettors. BRAND™ Rack Packed Tubes are ideal for biological tests, such as PCR*, cell uptake studies, RIA and EIA.
Have Questions?
Catalog NumberPackaging
FKS1012144196 Individual Tubes x 10 Units
Catalog number FKS10121441
Price (MYR)
1,346.00
Case
Packaging:
96 Individual Tubes x 10 Units
Price (MYR)
1,346.00
Case
  • Polypropylene tubes and rack are autoclavable at 121°C for 20min (except polyethylene caps)
  • Compatible with standard microplates and suitable for working with multi-channel pipettors
  • Size of tubes: outer diameter 8.8mm, height 45mm Ideal for biological tests, such as PCR*, cell uptake studies, RIA and EIA.
Specifications
Closure MaterialPolyethylene
AutoclavableAutoclavable
Capacity (Metric)1.2 mL
MaterialPolypropylene
For Use With (Application)Ideal for biological tests, such as PCR, cell uptake studies, RIA and EIA.
Height (Metric)45mm
No. per Pack960Pack
Packaging96 Individual Tubes x 10 Units
Temperature Range (Metric)121°C
SterilityNon-sterile
Diameter (Metric)8.8 mm
Unit SizeCase