(TPX™) PMP Low Form Measuring Cylinders
(TPX™) PMP Low Form Measuring Cylinders
Actual product may vary
Fisherbrand
(TPX™) PMP Low Form Measuring Cylinders
Accurately measures the volume of a liquid. Kartell™ Low Form Measuring Cylinders use horizontal marked lines to assist in measuring out various volumes of a liquid.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS10080971100 mL25 mL
Catalog number FKS10080971
Price (MYR)
567.00
PK
Add to cart
Capacity (Metric):
100 mL
Graduations:
25 mL
Price (MYR)
567.00
PK
Add to cart
Accurately measures the volume of a liquid. Kartell™ Low Form Measuring Cylinders use horizontal marked lines to assist in measuring out various volumes of a liquid.
  • Low form
  • Circular base
  • Good chemical resistance
  • Permanent graduations moulded into cylinders
  • Can withstand temperatures up to 170°C
  • Suitable for applications with food
Specifications
Capacity (Metric)100 mL
MaterialPMP
Tolerance±5 mL
No. per Pack2Pack
ScalePermanent Graduations Molded
Diameter (Metric)34.5 mm
Graduations25 mL
AutoclavableAutoclavable
Max. Temperature170°C
Subdivision5mL
ShapeCylindrical
For Use With (Application)Foodstuff
Base TypeCircular
Height (English)6.96 in.
Height (Metric)177 mm
TypeGraduated Measuring Cylinder
Unit SizePK