Nunc™ Cell Culture Tubes
Actual product may vary
Fisherbrand
Nunc™ Cell Culture Tubes
Perform adherent cell cultures in biotechnology, pharmaceutical and research laboratories with Thermo Scientific™ Nunc™ Cell Culture Tubes.
Have Questions?
Catalog NumberClosure TypeVolume (Metric) WorkingShape
FKS10048960Screw Cap3 mLFlat
Catalog number FKS10048960
Price (MYR)
6,339.00
Pack
Closure Type:
Screw Cap
Volume (Metric) Working:
3 mL
Shape:
Flat
Price (MYR)
6,339.00
Pack
Perform adherent cell cultures in biotechnology, pharmaceutical and research laboratories, as well as in the production of vaccines and diagnostic kits with Thermo Scientific™ Nunc™ Cell Culture Tubes.
Features:
  • Polystyrene tubes, polyethylene caps
  • Flat-sided tube provides 5.5cm2 culture area for grow of adherent cells
  • Flat-sided tube allows easy microscopy; a coverslip can be used in the tube
  • Medium retained in the flat-sided tube in horizontal position
  • Treated with Thermo Scientific™ Nunclon™ Delta covering
Warranty: 90 days.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure MaterialPolyethylene
Diameter (Metric)16 mm
Product TypeCulture Tube
Closure ColorWhite
DescriptionCell Culture Tube, 3mL
Closure TypeScrew Cap
ColorNatural
Length (Metric)110 mm
MaterialPolystyrene
No. per Pack75
No. per Case450
Volume (Metric) Working3 mL
Surface TreatmentNunclon Delta Cell Culture Treated
SterilitySterile
ShapeFlat
Unit SizePack