▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Potassium Dihydrogen Orthophosphate, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Potassium Dihydrogen Orthophosphate, Certified AR for Analysis, Fisher Chemical™
Potassium Dihydrogen Orthophosphate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 7778-77-0, White, Melting Point: 253 deg.C, Molecular Formula: H2KO4P, Molecular Weight: 136.084, Packaging: Plastic powder jar, pH: 4.5
Have Questions?
Change view
Quantity:
1 kg
250 g
500 g
3 kg
Catalog number
FKCP480060
also known as FKCP/4800/60
Price (MYR)
295.00
Bottle
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
295.00
Bottle
Add to cart
Specifications
Melting Point
253°C
CAS
7778-77-0
Color
White
pH
4.5
Physical Form
Solid
View more
Potassium Dihydrogen Orthophosphate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 7778-77-0, White, Melting Point: 253 deg.C, Molecular Formula: H2KO4P, Molecular Weight: 136.084, Packaging: Plastic powder jar, pH: 4.5