ASTM Low Form Glass Beaker
ASTM Low Form Glass Beaker
Actual product may vary
ASTM Low Form Glass Beaker
Made from high quality borosilicate 3.3 glass. Extremely durable, with a low temperature gradient, and superior chemical resistance. Features a spout for easy pouring and graduations as per ASTM-E960.
Have Questions?
Catalog NumberCapacity (Metric)No. per Pack
FISS40880600 mL1
Catalog number FISS40880
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
600 mL
No. per Pack:
1
  • Made from high quality borosilicate 3.3 glass
  • White graduations for easy visibility
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Superior chemical durability
  • Low form
  • Autoclavable at 121°C
  • In compliance with DIN 12231 and ISO 3819 standards
  • Graduated per ASTM-E960
  • Specifications
    Capacity (English)20.28 oz.
    ColorClear
    DescriptionChemical durability
    Diameter (English) Outer3.54 in.
    Diameter (Metric) Inner87.5 mm
    Diameter (Metric) Outer90 mm
    Capacity (Metric)600 mL
    MaterialBorosilicate Glass
    For Use With (Application)Holding liquids
    Height (English)5.3 in.
    Height (Metric)12.7 cm
    Length (Metric)8.9 cm
    No. per Pack1
    Quantity1/Ea.
    TypeGlass Beakers
    Width (English)3.5 in.
    Width (Metric)8.9 cm
    SterilityNon-sterile
    AutoclavableAutoclavable
    DisposableReusable
    GraduatedYes
    Certifications/ComplianceASTM
    ScaleWhite Graduated Scale
    StyleLow Form
    Unit SizeEA