Disposable Graduated Transfer Pipettes
Disposable Graduated Transfer Pipettes
Actual product may vary
Disposable Graduated Transfer Pipettes
Ideal for hematology, urinalysis, chemistry and blood banking — anywhere there is a need for quick, safe transfer of fluids.
Have Questions?
Catalog number FISS05014
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel. Molded in one piece from unbreakable, see-through low-density polyethylene to eliminate the hazards of broken glass and exposure to infectious materials.
  • Available gamma radiation-sterilized or nonsterile
  • 5.8mL capacity with 0.25 mL graduations
  • Draw 3.4 mL per squeeze
  • Inert to biological fluids and most acids
  • One squeeze draws from 2.0 to 3.3 mL (depending on style used)
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)
Specifications
Bulb Draw (Metric)3.4 mL
MaterialPolyethylene
SterilityNon-sterile
GraduatedYes
Volume (Metric)5.8 mL
Length (Metric)15.4 cm