▸
Education Supplies
▸
General Classroom Products
▸
Magnifiers
▸
DeskBrite™ Magnifier Desk Lamp
Actual product may vary
Fisherbrand
DeskBrite™ Magnifier Desk Lamp
Flexible neck allows you to position magnifier where you need it.
Have Questions?
Catalog Number
Magnification
FISS01926
2, 5x
Catalog number
FISS01926
Price (MYR)
-
Magnification:
2, 5x
2X LED magnifier lamp with 5X power spot lens
Two super-bright LED lights Large, 4 in. (10.2cm) crystal-clear acrylic lens provides large viewing area
Plug into standard outlet or use three AA batteries (not included)
Magnification: 2X/5X Lens size: 4.3 in. (110mm) Weight: 1.2 lb. (0.5kg)
Dimensions: 7 x 5.75 x 20.5 in. (17.8 x 14.6 x 52.1cm)
Specifications
Lighting
LED
Weight (English)
19.2 oz.
Weight (Metric)
544.31 g
Diameter (Englis) Lens
4.3 in.
Diameter (Metric) Lens
10.92 cm
Magnification
2, 5x
Material
Acrylic
Product Type
Magnifier, Deskbrite Desk Lamp