Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Microscope Slides
▸
Microscope Slide Storage
▸
Microscopy Slide Holders
▸
5-Place Slide Mailer
Actual product may vary
Fisherbrand
5-Place Slide Mailer
Safe and secure transport of standard microscope slides
Have Questions?
Catalog Number
Color
Dimensions (L x W x H)
FISHS15986
Natural
1.7 x 0.9 x 3.5 in.
Catalog number
FISHS15986
Price (MYR)
65.00
Pack
Add to cart
Color:
Natural
Dimensions (L x W x H):
1.7 x 0.9 x 3.5 in.
Price (MYR)
65.00
Pack
Add to cart
Polypropylene container holds five standard slides (76 x 26 mm and 3 x 1 in.)
Inside wall features slotted channels to keep slides separated and to make removal easier.
Lid closes securely to keep slides safe.
Specifications
Type
Slide Mailer
Slide Capacity
5
Closure Type
Snap-top
Color
Natural
Dimensions (L x W x H)
1.7 x 0.9 x 3.5 in.
For Use With (Equipment)
Standard Slides
Material
Polypropylene
Quantity
25 Pk.