HDPE Coated Wire Racks
HDPE Coated Wire Racks
Actual product may vary
HDPE Coated Wire Racks
HDPE Coated Wire Racks
HDPE Coated Wire Racks
Fisher Catalogue
HDPE Coated Wire Racks
Chemically-resistant and durable for transport or sample handling. HDPE Coated Wire Racks suitable for use with conical, round, or flat-bottom plastic tubes and bottles.
Have Questions?
Catalog NumberPositionsDiameter (Metric) Well
FISFB147916A825 to 30 mm
Catalog number FISFB147916A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Positions:
8
Diameter (Metric) Well:
25 to 30 mm
  • HDPE Coated Wire Racks
  • Chemically-resistant
  • Holds all brands and types of conical, round or flat-bottom, glass or plastic tubes and/or bottles
  • Square wells and three deck levels keep tubes secure
  • Use between temperatures of -40°C and 121°C
  • Slotted Bottom Support
  • Autoclavable
Specifications
Product TypeTube Rack
Array2 x 4
AutoclavableAutoclavable
ColorBlue
HoldsTubes
MaterialHDPE-coated Wire
Positions8
StackableNo
SterilityNon-sterile
No. per Case96/Cs.
Dimensions (L x W x H)5.8 x 3.20 x 3.50 in.
Diameter (Metric) Well25 to 30 mm
Weight (English)0.3 lb.
Weight (Metric)0.14 kg
Unit SizeEA