Disposable PES Filter Units
Disposable PES Filter Units
Actual product may vary
Disposable PES Filter Units
Disposable PES Filter Units
Fisherbrand
Disposable PES Filter Units
Increase cell culture sample protection with Fisherbrand sterile disposable vacuum filter units with PES membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeVolume (Metric)Diameter (Metric) MembraneFor Use With (Application)
FISFB125665060.20 μm1000 mL90 mmSterile Filtration of media and serum
FISFB125665010.45 μm150 mL50 mmFluid clarification and particle removal
FISFB125665020.20 μm250 mL50 mmSterile Filtration of media and serum
FISFB125665030.45 μm250 mL50 mmFluid clarification and particle removal
FISFB125665040.20 μm500 mL75 mmSterile Filtration of media and serum
Catalog number FISFB12566506
Price (MYR)
1,570.00
EA
Add to cart
Pore Size:
0.20 μm
Volume (Metric):
1000 mL
Diameter (Metric) Membrane:
90 mm
For Use With (Application):
Sterile Filtration of media and serum
Price (MYR)
1,570.00
EA
Add to cart
Increase cell culture sample protection with Fisherbrand sterile disposable vacuum filter units with PES membranes. The polyethersulfone (PES) membranes have low extractables and low protein binding for use in sterile filtration and clarification of cell culture media and serum.
  • PES membrane is low protein binding to maintain protein balance
  • PES membrane has low extractables to maintain media purity
  • Upper chamber of filter units is clearly marked with pore size, membrane type, catalog number, lot number and expiration date
  • Non-pyrogenic and non-cytotoxic
  • Gamma radiation-sterilized and individually bagged for a 5-year sterile shelf life
  • 0.2μm units for sterile filtration
  • 0.45μm units for fluid clarification and particle removal
Specifications
Binding CapacityLow binding
Certifications/ComplianceNon-pyrogenic per USP Standard Lot-to-Lot
DisposableSingle-use
MaterialPES
Material (Membrane)PES
Pore Size0.20 μm
SterilitySterile
For Use With (Application)Sterile Filtration of media and serum
TypeFilter Unit
Volume (Metric)1000 mL
Diameter (Metric) Membrane90 mm
Unit SizeEA