Short-Length Microcaps™ Micropipets
Actual product may vary
Fisherbrand
Short-Length Microcaps™ Micropipets
Ideal for handling dangerous materials
Have Questions?
Catalog NumberVolume (Metric)Length (Metric)
FIS21-170N100 μL116 mm
Catalog number FIS21-170N
Price (MYR)
-
Volume (Metric):
100 μL
Length (Metric):
116 mm
Ideal for handling dangerous materials
Precision-made capillary glass tubing, accurate to within ±1%.
  • To use, insert pipet through cap on end of rubber bulb assembly until visible through glass tube
  • Hole in rubber bulb permits free air circulation, fills pipet by capillary action
  • Pipet contains stated volume when completely filled from end to end
  • To discharge, hold finger over hole in bulb and squeeze
  • To rinse, repeat operation with diluent
Specifications
Accuracy±1%
CalibrationTo Contain
Diameter (English) Inner0.0413 in.
Diameter (English) Outer0.0565 in.
Diameter (Metric) Inner0.79 mm
Diameter (Metric) Outer1.43 mm
MaterialGlass
For Use With (Application)For handling dangerous materials, fast, safe, clean and accurate way to transfer liquids in micro quantities
IncludesBulb assembly
Length (English)4.56 in.
Volume (Metric)100 μL
Length (Metric)116 mm
TypeMicrocaps™ Micropipet