Barricade Tapes
Barricade Tapes
Actual product may vary
Harris Industries
Barricade Tapes
Increase visibility of hazards to help prevent accidents
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (L x W)Displayed Text
FIS19-019972Red1000 ft. x 3 in.Danger Danger
FIS19-019973Red1000 ft. x 3 in.Danger Do Not Enter
FIS19-019-948Yellow1000 ft. x 3 in.Caution Caution Caution
Catalog number FIS19-019972
Price (MYR)
-
Color:
Red
Dimensions (L x W):
1000 ft. x 3 in.
Displayed Text:
Danger Danger
Strong, lightweight polyethylene tapes with continuous repetition of message.
  • Can be tied, stapled, or nailed into place
  • Economical and reusable
  • Caution tapes have yellow background
  • Danger tapes have red background
Specifications
Certifications/ComplianceOSHA 1910.144
Dimensions (L x W)1000 ft. x 3 in.
Displayed TextDanger Danger
For Use With (Application)Increase visibility of hazards to help prevent accidents
Length (English)1000 ft.
Length (Metric)304.8 m
Thickness4 mil
Width (English)3 in.
Width (Metric)7.62 cm
Adhesive TypeNon-adhesive
ColorRed
MaterialHDPE
TypeBarricade Tape
Unit SizeCase