▸
Specialty Lab Equipment, Instruments, and Apparatuses
▸
General Purpose Forceps and Tweezers
▸
General-Purpose Tweezers with Straight Tips
Actual product may vary
Excelta
General-Purpose Tweezers with Straight Tips
General-Purpose Tweezers with Straight Tips feature a serrated design for better grip and handling.
Have Questions?
Catalog Number
Length (English)
Tip Style
Material
FIS17-467-234
6 in.
Straight, Blunt, Serrated
Stainless Steel
Catalog number
FIS17-467-234
Price (MYR)
100.00
Each
Add to cart
Length (English):
6 in.
Tip Style:
Straight, Blunt, Serrated
Material:
Stainless Steel
Price (MYR)
100.00
Each
Add to cart
Anti-magnetic stainless-steel
Straight broad serrated tips
High concentration of molybdenum provides high resistance to many acids
Specifications
Product Type
Tweezers, General Purpose
Length (English)
6 in.
Length (Metric)
152 mm
Material
Stainless Steel
Quantity
1 Ea.
Style
Jeweler
Tip Style
Straight, Blunt, Serrated
Width (English)
0.41 in.
Width (Metric)
1.04 cm