Enterprise Services
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Service Support Plans
Instrument Services
New Lab Project Services
Documents and Certificates
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Filters and Filtration
▸
Filtration Membrane Discs
▸
Filter Disc, Untreated, Ground Cylindrical, 20 mm
Actual product may vary
Fisherbrand
Filter Disc, Untreated, Ground Cylindrical, 20 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Have Questions?
Catalog Number
Porosity
FIS15-200
0
Catalog number
FIS15-200
Price (MYR)
53.00
Each
Add to cart
Porosity:
0
Price (MYR)
53.00
Each
Add to cart
Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Material
Borosilicate Glass (3.3)
Sterility
Non-sterile
No. per Pack
10/Pk.
Porosity
0
Thickness (Metric)
3.0 ± 0.7 mm
Diameter (Metric)
20 mm