▸
Humidity and Hygrometry
▸
Hydrometers
▸
Specific Gravity/Baume Hydrometers
Actual product may vary
Fisherbrand
Specific Gravity/Baume Hydrometers
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
Have Questions?
Catalog Number
Certifications/Compliance
For Use With (Equipment)
Specific Gravity Range
Volume (Metric)
FIS11-542D
NIST, ASTM E100, ASTM E126
300mL Borosilicate Glass cylinder
1.000 to 2.000
140 mL
Catalog number
FIS11-542D
Price (MYR)
107.00
Each
Add to cart
Certifications/Compliance:
NIST, ASTM E100, ASTM E126
For Use With (Equipment):
300mL Borosilicate Glass cylinder
Specific Gravity Range:
1.000 to 2.000
Volume (Metric):
140 mL
Price (MYR)
107.00
Each
Add to cart
Fisherbrand™ Specific Gravity/Baume Hydrometers are calibrated according to ASTM™ E 126 specifications.
Permanently marked with individual serial numbers
Specifications
Type
Specific Gravity Hydrometer
Increments
0.02
Certifications/Compliance
NIST, ASTM E100, ASTM E126
Length (Metric)
165 mm
For Use With (Equipment)
300mL Borosilicate Glass cylinder
Material
Soda Lime Glass
For Use With (Application)
Measure the specific gravity of liquids heavier than WaterMeasure the specific gravity of liquids heavier than Water
Specific Gravity Range
1.000 to 2.000
Volume (Metric)
140 mL