Glass Beaker with Cover
Actual product may vary
Glass Beaker with Cover
The Fisherbrand™ Glass Beaker with Cover is an ideal accessory for the Fisherbrand™ 11211 ultrasonic cleaner. This is a safe and economical choice for any laboratory.
Have Questions?
Catalog NumberCapacity (English)Capacity (Metric)Type
FIS104600920.28 oz.0.6 LBeaker
Catalog number FIS1046009
Price (MYR)
-
Capacity (English):
20.28 oz.
Capacity (Metric):
0.6 L
Type:
Beaker
  • Clear glass for easy viewing
  • Stable design prevents tipping
  • Easy to open and close lid
  • Graduated design
Specifications
TypeBeaker
Capacity (Metric)0.6 L
Capacity (English)20.28 oz.
For Use With (Equipment)FB11211 Ultrasonic Cleaning Unit
IncludesLid
MaterialGlass
Unit SizePack