PYREX™ Class A Lifetime Red Volumetric Flask with Glass Standard Taper Stopper
Actual product may vary
Corning
PYREX™ Class A Lifetime Red Volumetric Flask with Glass Standard Taper Stopper
Sharp, clear graduation lines
Have Questions?
Catalog NumberCapacity (Metric)
FIS10-210-8A25 mL
Catalog number FIS10-210-8A
Price (MYR)
2,016.00
Pack
Capacity (Metric):
25 mL
Price (MYR)
2,016.00
Pack
The capacity mark is a fine line etched through a narrow red band into clear glass, making a filler unnecessary and giving high legibility, accuracy, and permanence. With standard taper stopper. Calibrated 'To Contain.'
For stopper only, see Cat. No. 7650.
Specifications
CalibrationTo Contain
Diameter (English) Outer1.57 in.
Capacity (Metric)25 mL
MaterialGlass, Borosilicate Glass
Height (English)4.8 in.
Height (Metric)12.2 cm
Neck StyleLong
No. per Case12/Cs.
No. per Pack6/Pk.
Product TypeVolumetric Flask
Tolerance±0.03 mL
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Mouth StyleJoint
SterilityNon-sterile
ClassA
Bottom ShapeFlat
Diameter (Metric) Outer40 mm
Closure Size9
Unit SizePack